Assay Details
| Gene Symbol: Park7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020569.3 | NP_065594.2 | 5 - 6 | 478 | 64 | |
| GenBank mRNA | AB015652.1 | - | 5 - 6 | 425 | 64 | |
| AK146368.1 | - | 5 - 6 | 390 | 64 | ||
| AK153948.1 | - | 5 - 6 | 435 | 64 | ||
| AK168341.1 | - | 5 - 6 | 408 | 64 | ||
| BC002187.1 | - | 5 - 6 | 384 | 3487763 | 64 | |
| CD776363.1 | - | 2 - 3 | 64 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Park7
Genomic Map