Assay Details
| Gene Symbol: Otor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020595.2 | NP_065620.1 | 3 - 4 | 369 | 90 | |
| GenBank mRNA | AF243504.1 | - | 3 - 4 | 369 | 90 | |
| AJ243939.1 | - | 3 - 4 | 384 | 90 | ||
| BC139152.1 | - | 3 - 4 | 468 | 8862174 | 90 | |
| BC139153.1 | - | 3 - 4 | 468 | 8862175 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205524 | BC139153.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205524 | AF243504.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205524 | AJ243939.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205524 | AK047965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205524 | BC139152.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205524 | BC139153.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205524 | AF243504.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205524 | AJ243939.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205524 | AK047965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205524 | BC139152.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419178 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143961 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205524 | BC139153.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205524 | AF243504.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205524 | AJ243939.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205524 | AK047965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205524 | BC139152.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205524 | BC139153.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205524 | AF243504.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205524 | AJ243939.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205524 | AK047965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205524 | BC139152.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21699927 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143961 | - | - |
| Whole Mouse Genome G4122A | A_51_P205524 | BC139153.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205524 | AF243504.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205524 | AJ243939.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205524 | AK047965.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205524 | BC139152.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205524 | BC139153.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205524 | AF243504.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205524 | AJ243939.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205524 | AK047965.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205524 | BC139152.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121469 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143961 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01646095 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01646087 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01403300 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01646097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21699986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02109551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01668522 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699998 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01306957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01495739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02109546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01355465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01646093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01646087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01890058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01245209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02262389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01495621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01151230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01495737 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01495620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01577188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01890055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01495616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01994088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21699995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01938600 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10476632 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476630 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476631 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476629 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425083_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10476628 | - | - |
| Mouse Genome 430 2.0 Array | 1425083_at | - | - |
| Mouse Genome 430 2.0 Array | 1425083_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57329_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57329_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200016798.mm.1 | NM_020595.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200016798.mm.1 | TC0200002128.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200016799.mm.1 | NM_020595.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200016799.mm.1 | TC0200002128.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425083_PM_at | - | - |
| Mouse Expression Set 430 | 1425083_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425083_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425083_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425083_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425083_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 567036 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3440010 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3440010 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003440010 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2652615 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2652615 | - | - |
| mouse-6 v1.1 (Illumina) | scl20209.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3440010 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20209.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440010 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2652615 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2652615 | - | - |

Otor
Genomic Map