Assay Details
| Gene Symbol: Slc46a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021053.4 | NP_066395.3 | 1 - 2 | 1270 | 77 | |
| GenBank mRNA | AF148145.1 | - | 1 - 2 | 1242 | 77 | |
| AK014784.1 | - | 1 - 2 | 1251 | 77 | ||
| AK017363.1 | - | 1 - 2 | 1270 | 77 | ||
| AK040063.1 | - | 1271 | 77 | |||
| BC140962.1 | - | 1 - 2 | 1203 | 9055574 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc46a2
Genomic Map