Assay Details
| Gene Symbol: Usp29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290994.1 | NP_001277923.1 | 8 - 9 | 1457 | 92 | |
| NM_021323.3 | NP_067298.2 | 7 - 8 | 1393 | 92 | ||
| GenBank mRNA | AF229257.1 | - | 8 - 9 | 1457 | 92 | |
| AK038752.1 | - | 7 - 8 | 1236 | 92 | ||
| EU233992.1 | - | 4 - 5 | 734 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF229257.1, AK038752.1, EU233992.1, NM_001290994.1, NM_021323.3 transcripts.
Set Membership: |
|

Usp29
Genomic Map