Assay Details
| Gene Symbol: Ttyh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB162929.1 | - | 7 - 8 | 889 | 91 | |
| AF190699.1 | - | 7 - 8 | 961 | 91 | ||
| AF190991.1 | - | 7 - 8 | 961 | 91 | ||
| AK015486.1 | - | 6 - 7 | 612 | 91 | ||
| AK018148.1 | - | 7 - 8 | 953 | 91 | ||
| AK031550.1 | - | 7 - 8 | 989 | 91 | ||
| AK042758.1 | - | 7 - 8 | 742 | 91 | ||
| AK043998.1 | - | 7 - 8 | 1009 | 91 | ||
| AK048413.1 | - | 7 - 8 | 960 | 91 | ||
| AK049903.1 | - | 6 - 7 | 695 | 91 | ||
| AK081981.1 | - | 7 - 8 | 963 | 91 | ||
| AK083126.1 | - | 6 - 7 | 1086 | 91 | ||
| BC046310.1 | - | 7 - 8 | 947 | 6307420 | 91 | |
| BC065694.1 | - | 7 - 8 | 953 | 6813365 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttyh1
Genomic Map