Assay Details
| Gene Symbol: Ttyh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB162929.1 | - | 10 - 11 | 1126 | 114 | |
| AF190699.1 | - | 10 - 11 | 1198 | 114 | ||
| AF190991.1 | - | 10 - 11 | 1198 | 114 | ||
| AK015486.1 | - | 9 - 10 | 849 | 114 | ||
| AK018148.1 | - | 10 - 11 | 1190 | 114 | ||
| AK031550.1 | - | 10 - 11 | 1226 | 114 | ||
| AK042758.1 | - | 10 - 11 | 979 | 114 | ||
| AK043998.1 | - | 10 - 11 | 1246 | 114 | ||
| AK048413.1 | - | 10 - 11 | 1197 | 114 | ||
| AK049903.1 | - | 9 - 10 | 932 | 114 | ||
| AK081981.1 | - | 10 - 11 | 1200 | 114 | ||
| AK083126.1 | - | 9 - 10 | 1323 | 114 | ||
| BC046310.1 | - | 10 - 11 | 1184 | 6307420 | 114 | |
| BC065694.1 | - | 10 - 11 | 1190 | 6813365 | 114 | |
More Information
Set Membership: |
|

Ttyh1
Genomic Map