Assay Details
| Gene Symbol: Ttyh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF190699.1 | - | 11 - 12 | 1344 | 55 | |
| AF190991.1 | - | 11 - 12 | 1344 | 55 | ||
| AK018148.1 | - | 11 - 12 | 1336 | 55 | ||
| AK042758.1 | - | 11 - 12 | 1125 | 55 | ||
| AK043998.1 | - | 11 - 12 | 1392 | 55 | ||
| AK048413.1 | - | 11 - 12 | 1343 | 55 | ||
| AK049903.1 | - | 10 - 11 | 1078 | 55 | ||
| AK081981.1 | - | 11 - 12 | 1346 | 55 | ||
| AK083126.1 | - | 10 - 11 | 1469 | 55 | ||
| BC046310.1 | - | 11 - 12 | 1330 | 6307420 | 55 | |
More Information
Set Membership: |
|

Ttyh1
Genomic Map