Assay Details
| Gene Symbol: Itgax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021334.2 | NP_067309.1 | 15 - 16 | 1891 | 73 | |
| GenBank mRNA | AF211864.1 | - | 15 - 16 | 1891 | 73 | |
| AK131133.1 | - | 15 - 16 | 1897 | 73 | ||
| AK155816.1 | - | 15 - 16 | 1913 | 73 | ||
| AK170432.1 | - | 15 - 16 | 1909 | 73 | ||
| BC042840.1 | - | 15 - 16 | 1890 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itgax
Genomic Map