Assay Details
| Gene Symbol: Gsdma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021347.4 | NP_067322.1 | 7 - 8 | 822 | 78 | |
| GenBank mRNA | AB033595.1 | - | 7 - 8 | 811 | 78 | |
| AK028645.1 | - | 7 - 8 | 823 | 78 | ||
| AK028698.1 | - | 7 - 8 | 823 | 78 | ||
| AK029062.1 | - | 7 - 8 | 806 | 78 | ||
More Information
Set Membership: |
|

Gsdma
Genomic Map