Assay Details
Gene Symbol: Tslp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021367.2 | NP_067342.1 | 3 - 4 | 198 | 91 | |
GenBank mRNA | AF232937.1 | - | 3 - 4 | 198 | 91 | |
BC130230.1 | - | 3 - 4 | 202 | 40141922 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
