Assay Details
| Gene Symbol: Shank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021423.3 | NP_067398.2 | 13 - 14 | 1685 | 82 | |
| GenBank mRNA | AB231013.1 | - | 13 - 14 | 1685 | 82 | |
| AB841411.1 | - | 3 - 4 | 649 | 82 | ||
| AB841412.1 | - | 3 - 4 | 649 | 82 | ||
| AJ245904.1 | - | 11 - 12 | 1539 | 82 | ||
| HQ405759.1 | - | 1 - 2 | 116 | 82 | ||
| HQ405760.1 | - | 1 - 2 | 116 | 82 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the HQ405760.1 transcripts.
Set Membership: |
|

Shank3
Genomic Map