Assay Details
| Gene Symbol: Taf1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021441.2 | NP_067416.2 | 9 - 10 | 1070 | 83 | |
| GenBank mRNA | AK030522.1 | - | 9 - 10 | 1037 | 83 | |
| AK052250.1 | - | 9 - 10 | 1110 | 83 | ||
| AK087534.1 | - | 9 - 10 | 1060 | 83 | ||
| AK154846.1 | - | 9 - 10 | 1054 | 83 | ||
| AK156718.1 | - | 9 - 10 | 1037 | 83 | ||
| BC051944.1 | - | 9 - 10 | 1122 | 83 | ||
| BC058383.1 | - | 9 - 10 | 1012 | 6811470 | 83 | |
| Y09974.1 | - | 9 - 10 | 1197 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taf1c
Genomic Map