| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600013745.mm.1 |
NM_021496.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013745.mm.1 |
NM_021496.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013745.mm.1 |
TC1600001679.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600013751.mm.1 |
NM_021496.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013751.mm.1 |
NM_021496.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013751.mm.1 |
NM_021497.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013751.mm.1 |
NM_021497.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013751.mm.1 |
TC1600001679.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600013752.mm.1 |
NM_021496.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013752.mm.1 |
NM_021496.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013752.mm.1 |
NM_021497.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013752.mm.1 |
NM_021497.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013752.mm.1 |
TC1600001679.mm.1 |
MTA TC |