Assay Details
| Gene Symbol: Wrap73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021499.2 | NP_067474.2 | 9 - 10 | 989 | 75 | |
| GenBank mRNA | AB034911.2 | - | 9 - 10 | 969 | 75 | |
| AK011773.1 | - | 9 - 10 | 952 | 75 | ||
| AK051688.1 | - | 9 - 10 | 933 | 75 | ||
| AK156969.1 | - | 9 - 10 | 988 | 75 | ||
| AK160331.1 | - | 9 - 10 | 961 | 75 | ||
| AK172690.1 | - | 9 - 10 | 988 | 75 | ||
| BC016120.1 | - | 8 - 9 | 2616 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wrap73
Genomic Map