Assay Details
| Gene Symbol: Trpv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022413.4 | NP_071858.3 | 8 - 9 | 1417 | 70 | |
| GenBank mRNA | AB037373.1 | - | 8 - 9 | 1321 | 70 | |
| AJ542487.1 | - | 8 - 9 | 1122 | 70 | ||
| AK077234.1 | - | 8 - 9 | 1347 | 70 | ||
| AK085123.1 | - | 5 - 6 | 2107 | 70 | ||
| AK168760.1 | - | 8 - 9 | 1417 | 70 | ||
| BC016101.1 | - | 8 - 9 | 1355 | 4911355 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trpv6
Genomic Map