Assay Details
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020044.3 | NP_064428.1 | 1 - 2 | 137 | 116 | |
NM_022964.4 | NP_075253.2 | 1 - 2 | 137 | 116 | ||
GenBank mRNA | AK138953.1 | - | 1 - 2 | 96 | 116 | |
AK143533.1 | - | 1 - 2 | 138 | 116 | ||
AK144261.1 | - | 1 - 2 | 76 | 116 | ||
AK162321.1 | - | 1 - 2 | 96 | 116 | ||
AY190024.1 | - | 1 - 2 | 102 | 116 | ||
BY228290.1 | - | 1 - 2 | 136 | 116 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK138953.1,AK143533.1,AK162321.1,AY190024.1,NM_020044.3,NM_022964.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
