Assay Details
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020044.3 | NP_064428.1 | 2 - 3 | 259 | 100 | |
NM_022964.4 | NP_075253.2 | 2 - 3 | 259 | 100 | ||
GenBank mRNA | AF257136.1 | - | 1 - 2 | 203 | 100 | |
AK138953.1 | - | 2 - 3 | 218 | 100 | ||
AK143533.1 | - | 2 - 3 | 260 | 100 | ||
AK144261.1 | - | 2 - 3 | 198 | 100 | ||
AK162321.1 | - | 2 - 3 | 218 | 100 | ||
AY190024.1 | - | 2 - 3 | 224 | 100 | ||
BC005804.1 | - | 2 - 3 | 188 | 3154073 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
