Assay Details
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020044.3 | NP_064428.1 | 4 - 5 | 347 | 113 | |
NM_022964.4 | NP_075253.2 | 4 - 5 | 347 | 113 | ||
GenBank mRNA | AF257136.1 | - | 3 - 4 | 291 | 113 | |
AK134721.1 | - | 1 - 2 | 39 | 113 | ||
AK138953.1 | - | 4 - 5 | 306 | 113 | ||
AK143533.1 | - | 4 - 5 | 348 | 113 | ||
AK144261.1 | - | 4 - 5 | 286 | 113 | ||
AK162321.1 | - | 4 - 5 | 306 | 113 | ||
AY190024.1 | - | 4 - 5 | 312 | 113 | ||
BC005804.1 | - | 4 - 5 | 276 | 3154073 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
