Assay Details
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020044.3 | NP_064428.1 | 10 - 11 | 621 | 108 | |
NM_022964.4 | NP_075253.2 | 9 - 10 | 585 | 108 | ||
GenBank mRNA | AF257136.1 | - | 9 - 10 | 565 | 108 | |
AK134721.1 | - | 7 - 8 | 313 | 108 | ||
AK138953.1 | - | 10 - 11 | 580 | 108 | ||
AK143533.1 | - | 9 - 10 | 586 | 108 | ||
AK162321.1 | - | 9 - 10 | 544 | 108 | ||
AY190024.1 | - | 10 - 11 | 586 | 108 | ||
BC005804.1 | - | 9 - 10 | 514 | 3154073 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
