Assay Details
| Gene Symbol: Lrp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022993.3 | NP_075369.2 | 5 - 6 | 1854 | 85 | |
| GenBank mRNA | AB042200.1 | - | 5 - 6 | 1756 | 85 | |
| AK132169.1 | - | 1838 | 85 | |||
| AK144035.1 | - | 5 - 6 | 1867 | 85 | ||
| AK170325.1 | - | 5 - 6 | 1828 | 85 | ||
| BC011058.1 | - | 1 - 2 | 134 | 85 | ||
| BC052378.1 | - | 5 - 6 | 1830 | 5696275 | 85 | |
| BC054447.1 | - | 5 - 6 | 1822 | 6477698 | 85 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC011058.1 transcripts.
Set Membership: |
|

Lrp10
Genomic Map