| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200015940.mm.1 |
NM_001111075.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015940.mm.1 |
NM_023117.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015940.mm.1 |
TC0200002010.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015942.mm.1 |
NM_001111075.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015942.mm.1 |
NM_023117.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015942.mm.1 |
TC0200002010.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015941.mm.1 |
NM_001111075.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015941.mm.1 |
NM_023117.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015941.mm.1 |
TC0200002010.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015939.mm.1 |
NM_001111075.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015939.mm.1 |
NM_023117.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015939.mm.1 |
TC0200002010.mm.1 |
MTA TC |