Assay Details
| Gene Symbol: Cdc25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023117.4 | NP_075606.1 | 7 - 8 | 934 | 83 | |
| GenBank mRNA | AK153903.1 | - | 7 - 8 | 935 | 83 | |
| S93521.1 | - | 7 - 8 | 803 | 83 | ||
| W54021.1 | - | 3 - 4 | 83 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cdc25b
Genomic Map