Assay Details
| Gene Symbol: Ankra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271388.1 | NP_001258317.1 | 3 - 4 | 946 | 61 | |
| NM_001271389.1 | NP_001258318.1 | 3 - 4 | 946 | 61 | ||
| NM_001271390.1 | NP_001258319.1 | 2 - 3 | 564 | 61 | ||
| NM_001271391.1 | NP_001258320.1 | 2 - 3 | 564 | 61 | ||
| NM_001271392.1 | NP_001258321.1 | 2 - 3 | 564 | 61 | ||
| NM_023472.2 | NP_075961.1 | 3 - 4 | 946 | 61 | ||
| GenBank mRNA | AB221648.1 | - | 2 - 3 | 449 | 61 | |
| AF314031.1 | - | 3 - 4 | 780 | 61 | ||
| AK003438.2 | - | 2 - 3 | 483 | 61 | ||
| AK054037.1 | - | 3 - 4 | 751 | 61 | ||
| AK081695.1 | - | 2 - 3 | 449 | 61 | ||
| AK161446.1 | - | 3 - 4 | 950 | 61 | ||
| BC037437.1 | - | 2 - 3 | 492 | 61 | ||
| BC066113.1 | - | 2 - 3 | 483 | 30361246 | 61 | |
| BC079615.1 | - | 3 - 4 | 763 | 30361702 | 61 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK081695.1,NM_001271390.1,NM_001271391.1,NM_001271392.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29256605 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29256604 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29256607 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29256606 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114009 | BC066113.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114009 | BC037437.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114009 | AF314031.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114009 | NM_023472.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 865388 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621253 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 163443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419680 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188158 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 160070 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107574 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154511 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114009 | BC066113.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114009 | BC037437.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114009 | AF314031.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114009 | NM_023472.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P235557 | BC079615.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P235557 | BC028323.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P235557 | AK139343.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P235557 | AK161446.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P235557 | AF314031.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368804 | BC066113.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368804 | AF314031.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368804 | NM_023472.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29256608 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07967058 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107574 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154511 | - | - |
| Whole Mouse Genome G4122A | A_52_P360413 | AK171682.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P368804 | BC066113.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P368804 | AF314031.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P368804 | NM_023472.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P235557 | BC079615.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P235557 | BC028323.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P235557 | AK139343.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P235557 | AK161446.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P235557 | AF314031.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107655 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107574 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154511 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10751378 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10751374 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10751370 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10751379 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10751372 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29256606 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29256605 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29256604 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29256607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29256681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256648 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10829016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10560430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10518590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10826293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10751372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10953512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10588190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29256645 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10406828 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406832 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406836 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406829 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406830 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406837 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406835 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406827 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406834 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406833 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406831 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430668_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441935_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10406825 | - | - |
| Mouse Genome 430 2.0 Array | 1430668_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1430668_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1441935_at | - | - |
| Mouse Genome 430 2.0 Array | 1441935_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68558_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68558_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300007095.mm.1 | NM_001271388.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007095.mm.1 | NM_001271389.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007095.mm.1 | NM_001271390.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007095.mm.1 | NM_001271391.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007095.mm.1 | NM_001271392.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007095.mm.1 | NM_023472.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007095.mm.1 | TC1300001112.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430668_a_at | - | - |
| Mouse Expression Set 430 | 1430668_PM_a_at | - | - |
| Mouse Expression Set 430 | 1441935_at | - | - |
| Mouse Expression Set 430 | 1441935_PM_at | - | - |
| Murine Genome U74v2 Set | 167932_PM_f_at | - | - |
| Murine Genome U74v2 Set | 167932_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430668_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430668_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430668_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430668_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441935_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441935_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 556218 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102570204 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3060671 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104670132 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102570204 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3060671 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105550288 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003060671 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2660477 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257723 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2660477 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228841 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243090 | - | - |
| mouse-6 v1.1 (Illumina) | scl44477.7_25-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104670132 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102570204 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105550288 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3060671 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44477.7_25-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104670132 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3060671 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102570204 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105550288 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2660477 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1243090 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257723 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2660477 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2543304 | - | - |

Ankra2
Genomic Map