Assay Details
| Gene Symbol: Ankra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271388.1 | NP_001258317.1 | 6 - 7 | 1229 | 72 | |
| NM_001271389.1 | NP_001258318.1 | 6 - 7 | 1229 | 72 | ||
| NM_001271390.1 | NP_001258319.1 | 5 - 6 | 847 | 72 | ||
| NM_001271391.1 | NP_001258320.1 | 5 - 6 | 847 | 72 | ||
| NM_001271392.1 | NP_001258321.1 | 5 - 6 | 847 | 72 | ||
| NM_023472.2 | NP_075961.1 | 6 - 7 | 1229 | 72 | ||
| GenBank mRNA | AB221648.1 | - | 5 - 6 | 732 | 72 | |
| AF314031.1 | - | 6 - 7 | 1063 | 72 | ||
| AK003438.2 | - | 5 - 6 | 766 | 72 | ||
| AK054037.1 | - | 6 - 7 | 1035 | 72 | ||
| AK081695.1 | - | 5 - 6 | 732 | 72 | ||
| AK139343.1 | - | 6 - 7 | 1111 | 72 | ||
| AK161446.1 | - | 6 - 7 | 1235 | 72 | ||
| BC066113.1 | - | 5 - 6 | 766 | 30361246 | 72 | |
| BC079615.1 | - | 6 - 7 | 1046 | 30361702 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ankra2
Genomic Map