Assay Details
| Gene Symbol: Capn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023709.4 | NP_076198.2 | 8 - 9 | 985 | 83 | |
| GenBank mRNA | AK008629.1 | - | 8 - 9 | 986 | 83 | |
| AK144514.1 | - | 8 - 9 | 988 | 83 | ||
| BC058748.1 | - | 8 - 9 | 963 | 6394969 | 83 | |
| U89513.1 | - | 8 - 9 | 946 | 83 | ||
More Information
Set Membership: |
|

Capn9
Genomic Map