Assay Details
| Gene Symbol: Gmpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002326.1 | - | 6 - 7 | 843 | 65 | |
| AK009159.1 | - | 6 - 7 | 854 | 65 | ||
| AK084508.1 | - | 6 - 7 | 771 | 65 | ||
| BC010827.1 | - | 6 - 7 | 825 | 4219628 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmpr
Genomic Map