Assay Details
| Gene Symbol: Ergic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025516.4 | NP_079792.1 | 2 - 3 | 180 | 73 | |
| GenBank mRNA | AK009350.1 | - | 2 - 3 | 181 | 73 | |
| AK013942.1 | - | 2 - 3 | 180 | 73 | ||
| AK050285.1 | - | 2 - 3 | 195 | 73 | ||
| BC043720.1 | - | 2 - 3 | 205 | 5375947 | 73 | |
| BC057130.1 | - | 2 - 3 | 199 | 6819420 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ergic3
Genomic Map