Assay Details
| Gene Symbol: Taf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025579.3 | NP_079855.2 | 2 - 3 | 359 | 144 | |
| GenBank mRNA | AK007811.1 | - | 2 - 3 | 322 | 144 | |
| AK013133.1 | - | 2 - 3 | 322 | 144 | ||
| AK132927.1 | - | 2 - 3 | 360 | 144 | ||
| AK139748.1 | - | 2 - 3 | 273 | 144 | ||
| BC019668.1 | - | 2 - 3 | 304 | 3493047 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Taf12
Genomic Map