Assay Details
| Gene Symbol: Ctnnbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025680.4 | NP_079956.3 | 1 - 2 | 153 | 86 | |
| GenBank mRNA | AK010547.1 | - | 1 - 2 | 64 | 86 | |
| AK044690.1 | - | 1 - 2 | 93 | 86 | ||
| AK077616.1 | - | 1 - 2 | 97 | 86 | ||
| AK145147.1 | - | 1 - 2 | 105 | 86 | ||
| AK145598.1 | - | 1 - 2 | 97 | 86 | ||
| AK145719.1 | - | 1 - 2 | 97 | 86 | ||
| AK145888.1 | - | 1 - 2 | 114 | 86 | ||
| AK146274.1 | - | 1 - 2 | 113 | 86 | ||
| AY009405.1 | - | 1 - 2 | 113 | 86 | ||
| BC050787.1 | - | 1 - 2 | 123 | 6705648 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctnnbl1
Genomic Map