Assay Details
| Gene Symbol: Ctnnbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025680.4 | NP_079956.3 | 6 - 7 | 780 | 66 | |
| GenBank mRNA | AK010547.1 | - | 6 - 7 | 691 | 66 | |
| AK044690.1 | - | 6 - 7 | 720 | 66 | ||
| AK077616.1 | - | 6 - 7 | 724 | 66 | ||
| AK145147.1 | - | 6 - 7 | 735 | 66 | ||
| AK145598.1 | - | 6 - 7 | 724 | 66 | ||
| AK145719.1 | - | 6 - 7 | 724 | 66 | ||
| AK145888.1 | - | 6 - 7 | 741 | 66 | ||
| AK146274.1 | - | 6 - 7 | 740 | 66 | ||
| AY009405.1 | - | 6 - 7 | 740 | 66 | ||
| BC038332.1 | - | 10 - 11 | 66 | |||
| BC050787.1 | - | 6 - 7 | 750 | 6705648 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctnnbl1
Genomic Map