Assay Details
| Gene Symbol: Ctnnbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025680.4 | NP_079956.3 | 8 - 9 | 950 | 65 | |
| GenBank mRNA | AK010547.1 | - | 8 - 9 | 861 | 65 | |
| AK044690.1 | - | 8 - 9 | 890 | 65 | ||
| AK077616.1 | - | 8 - 9 | 894 | 65 | ||
| AK145147.1 | - | 8 - 9 | 906 | 65 | ||
| AK145598.1 | - | 8 - 9 | 894 | 65 | ||
| AK145719.1 | - | 8 - 9 | 894 | 65 | ||
| AK145888.1 | - | 8 - 9 | 911 | 65 | ||
| AK146274.1 | - | 8 - 9 | 910 | 65 | ||
| AY009405.1 | - | 8 - 9 | 910 | 65 | ||
| BC038332.1 | - | 8 - 9 | 65 | |||
| BC050787.1 | - | 8 - 9 | 920 | 6705648 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctnnbl1
Genomic Map