Assay Details
| Gene Symbol: Lztr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025808.3 | NP_080084.2 | 5 - 6 | 559 | 86 | |
| GenBank mRNA | AK004561.2 | - | 5 - 6 | 559 | 86 | |
| AK005037.1 | - | 5 - 6 | 558 | 86 | ||
| AK083411.1 | - | 5 - 6 | 563 | 86 | ||
| AK132817.1 | - | 5 - 6 | 558 | 86 | ||
| AK206763.1 | - | 1 - 2 | 115 | 86 | ||
| AK215859.1 | - | 1 - 2 | 119 | 86 | ||
| BC034400.1 | - | 5 - 6 | 556 | 2651456 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lztr1
Genomic Map