Assay Details
| Gene Symbol: Fbxo32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026346.3 | NP_080622.1 | 2 - 3 | 508 | 102 | |
| GenBank mRNA | AF441120.1 | - | 2 - 3 | 554 | 102 | |
| AK014612.1 | - | 2 - 3 | 461 | 102 | ||
| AK014793.1 | - | 2 - 3 | 490 | 102 | ||
| BC027211.1 | - | 2 - 3 | 442 | 3981597 | 102 | |
| BU921446.1 | - | 2 - 3 | 508 | 102 | ||
More Information
Set Membership: |
|

Fbxo32
Genomic Map