Assay Details
| Gene Symbol: Fbxo32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026346.3 | NP_080622.1 | 3 - 4 | 561 | 118 | |
| GenBank mRNA | AF441120.1 | - | 3 - 4 | 607 | 118 | |
| AK014612.1 | - | 3 - 4 | 514 | 118 | ||
| AK014793.1 | - | 3 - 4 | 543 | 118 | ||
| BC027211.1 | - | 3 - 4 | 495 | 3981597 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo32
Genomic Map