Assay Details
| Gene Symbol: Fbxo32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026346.3 | NP_080622.1 | 7 - 8 | 1112 | 71 | |
| GenBank mRNA | AF441120.1 | - | 7 - 8 | 1158 | 71 | |
| AK014612.1 | - | 7 - 8 | 1065 | 71 | ||
| AK014793.1 | - | 7 - 8 | 1094 | 71 | ||
| BC027211.1 | - | 7 - 8 | 1046 | 3981597 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo32
Genomic Map