Assay Details
| Gene Symbol: Cxxc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028868.3 | NP_083144.1 | 8 - 9 | 1303 | 62 | |
| GenBank mRNA | AK010337.1 | - | 8 - 9 | 1304 | 62 | |
| AK083655.1 | - | 8 - 9 | 1308 | 62 | ||
| AK146688.1 | - | 8 - 9 | 1387 | 62 | ||
| AY099096.1 | - | 8 - 9 | 1275 | 62 | ||
| BC030938.1 | - | 8 - 9 | 1135 | 3980339 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cxxc1
Genomic Map