Assay Details
| Gene Symbol: Spsb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF403036.1 | - | 1 - 2 | 235 | 92 | |
| AK015181.1 | - | 1 - 2 | 235 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF403036.1, AK015181.1 transcripts.
Set Membership: |
|

Spsb1
Genomic Map