Assay Details
| Gene Symbol: Dhx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030150.2 | NP_084426.2 | 1 - 2 | 99 | 83 | |
| GenBank mRNA | AK021012.1 | - | 1 - 2 | 99 | 83 | |
| BY727457.1 | - | 1 - 2 | 99 | 83 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_030150.2 transcripts.
Set Membership: |
|

Dhx58
Genomic Map