Assay Details
| Gene Symbol: Inpp5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290437.1 | NP_001277366.1 | 6 - 7 | 1639 | 81 | |
| NM_033134.3 | NP_149125.1 | 5 - 6 | 1923 | 81 | ||
| NR_110957.1 | 5 - 6 | 2010 | 81 | |||
| GenBank mRNA | AF226683.1 | - | 5 - 6 | 1885 | 81 | |
| AK080075.1 | - | 5 - 6 | 1905 | 81 | ||
| AK085439.1 | - | 6 - 7 | 1643 | 81 | ||
| AK154097.1 | - | 5 - 6 | 1924 | 81 | ||
| AK158135.1 | - | 5 - 6 | 1908 | 81 | ||
| AK170174.1 | - | 6 - 7 | 1929 | 81 | ||
| AK170786.1 | - | 5 - 6 | 1880 | 81 | ||
| AK170812.1 | - | 5 - 6 | 1894 | 81 | ||
| BC052717.1 | - | 5 - 6 | 1892 | 6837339 | 81 | |
| BC057566.1 | - | 5 - 6 | 1975 | 81 | ||
| BC080295.1 | - | 5 - 6 | 1888 | 30354397 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inpp5e
Genomic Map