Assay Details
| Gene Symbol: Inpp5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290437.1 | NP_001277366.1 | 8 - 9 | 1906 | 61 | |
| NM_033134.3 | NP_149125.1 | 7 - 8 | 2190 | 61 | ||
| NR_110957.1 | 7 - 8 | 2277 | 61 | |||
| GenBank mRNA | AF226683.1 | - | 7 - 8 | 2152 | 61 | |
| AK080075.1 | - | 7 - 8 | 2172 | 61 | ||
| AK085439.1 | - | 8 - 9 | 1911 | 61 | ||
| AK154097.1 | - | 7 - 8 | 2191 | 61 | ||
| AK158135.1 | - | 7 - 8 | 2175 | 61 | ||
| AK170786.1 | - | 7 - 8 | 2147 | 61 | ||
| BC052717.1 | - | 7 - 8 | 2159 | 6837339 | 61 | |
| BC057566.1 | - | 7 - 8 | 2242 | 61 | ||
| BC080295.1 | - | 7 - 8 | 2155 | 30354397 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inpp5e
Genomic Map