Assay Details
| Gene Symbol: Slc45a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053077.3 | NP_444307.1 | 3 - 4 | 979 | 75 | |
| GenBank mRNA | AF360357.1 | - | 3 - 4 | 887 | 75 | |
| AK029155.1 | - | 3 - 4 | 980 | 75 | ||
| AK089932.1 | - | 3 - 4 | 75 | |||
| AK148207.1 | - | 977 | 75 | |||
| AY034377.1 | - | 3 - 4 | 900 | 75 | ||
| BC125376.1 | - | 3 - 4 | 966 | 40129891 | 75 | |
| BC132429.1 | - | 3 - 4 | 966 | 40130706 | 75 | |
More Information
Set Membership: |
|

Slc45a2
Genomic Map