Assay Details
| Gene Symbol: Caskin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080643.2 | NP_542374.2 | 4 - 5 | 724 | 65 | |
| GenBank mRNA | AF451978.1 | - | 3 - 4 | 443 | 65 | |
| AK129297.1 | - | 4 - 5 | 762 | 65 | ||
| BC053083.1 | - | 4 - 5 | 724 | 5704642 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Caskin2
Genomic Map