Assay Details
| Gene Symbol: Caskin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080643.2 | NP_542374.2 | 14 - 15 | 1951 | 76 | |
| GenBank mRNA | AF451978.1 | - | 13 - 14 | 1670 | 76 | |
| AK037133.1 | - | 1 - 2 | 76 | 76 | ||
| AK129297.1 | - | 14 - 15 | 1989 | 76 | ||
| BC053083.1 | - | 14 - 15 | 1951 | 5704642 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Caskin2
Genomic Map