Assay Details
| Gene Symbol: Lnx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080795.4 | NP_542985.4 | 1 - 2 | 156 | 137 | |
| GenBank mRNA | AF401681.1 | - | 1 - 2 | 113 | 137 | |
| AK036218.1 | - | 1 - 2 | 157 | 137 | ||
| AK047053.1 | - | 1 - 2 | 140 | 138 | ||
| AK170146.1 | - | 1 - 2 | 110 | 137 | ||
| BC090665.1 | - | 1 - 2 | 122 | 30686384 | 137 | |
| BC117938.1 | - | 1 - 2 | 110 | 40097810 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF401681.1, AK036218.1, BC090665.1, BC117938.1, NM_080795.4 transcripts.
Set Membership: |
|

Lnx2
Genomic Map