Assay Details
| Gene Symbol: Lnx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080795.4 | NP_542985.4 | 6 - 7 | 1573 | 113 | |
| GenBank mRNA | AF401681.1 | - | 6 - 7 | 1530 | 113 | |
| AK036218.1 | - | 6 - 7 | 1574 | 113 | ||
| AK047053.1 | - | 6 - 7 | 1557 | 113 | ||
| AK140320.1 | - | 6 - 7 | 1508 | 113 | ||
| AK170146.1 | - | 6 - 7 | 1537 | 114 | ||
| BC090665.1 | - | 6 - 7 | 1539 | 30686384 | 113 | |
| BC117937.1 | - | 6 - 7 | 1528 | 40097802 | 113 | |
| BC117938.1 | - | 6 - 7 | 1527 | 40097810 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lnx2
Genomic Map