Assay Details
| Gene Symbol: Colec12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130449.2 | NP_569716.2 | 6 - 7 | 1918 | 129 | |
| GenBank mRNA | AB038519.1 | - | 6 - 7 | 1889 | 129 | |
| AB078434.1 | - | 6 - 7 | 1904 | 129 | ||
| AK042772.1 | - | 6 - 7 | 1919 | 129 | ||
| BC009162.1 | - | 6 - 7 | 129 | |||
| BC048915.1 | - | 2 - 3 | 604 | 129 | ||
| BC057936.1 | - | 6 - 7 | 1907 | 5326386 | 129 | |
| BC060657.1 | - | 6 - 7 | 1896 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Colec12
Genomic Map