Assay Details
| Gene Symbol: Recql5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130454.2 | NP_569721.1 | 2 - 3 | 823 | 71 | |
| GenBank mRNA | AB038773.1 | - | 2 - 3 | 157 | 71 | |
| AB059999.1 | - | 2 - 3 | 337 | 71 | ||
| AK083513.1 | - | 2 - 3 | 819 | 71 | ||
| BC023769.1 | - | 1 - 2 | 115 | 71 | ||
| BC053086.1 | - | 2 - 3 | 637 | 71 | ||
| BC059000.1 | - | 2 - 3 | 797 | 71 | ||
| BC060070.1 | - | 2 - 3 | 637 | 71 | ||
| BC066176.1 | - | 2 - 3 | 546 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Recql5
Genomic Map