Assay Details
| Gene Symbol: Cdhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130878.2 | NP_570948.1 | 11 - 12 | 1335 | 66 | |
| GenBank mRNA | AB093300.1 | - | 11 - 12 | 1751 | 66 | |
| AF426393.1 | - | 11 - 12 | 1222 | 66 | ||
| AK143151.1 | - | 11 - 12 | 1276 | 66 | ||
| BC042454.1 | - | 11 - 12 | 1245 | 5369576 | 66 | |
| BC058270.1 | - | 11 - 12 | 1335 | 6490597 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdhr1
Genomic Map