Assay Details
| Gene Symbol: Otud7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130880.1 | NP_570950.1 | 1 - 2 | 162 | 105 | |
| GenBank mRNA | AJ430384.1 | - | 1 - 2 | 162 | 105 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AJ430384.1,NM_130880.1 transcripts.
Set Membership: |
|

Otud7a
Genomic Map