Assay Details
| Gene Symbol: Scn4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133199.2 | NP_573462.2 | 3 - 4 | 486 | 82 | |
| GenBank mRNA | AJ278787.1 | - | 3 - 4 | 486 | 82 | |
| BC129805.1 | - | 3 - 4 | 484 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scn4a
Genomic Map